missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitroprusside Dihydrate, puriss. p.a., ≥99%, ACS reagent, Honeywell Fluka™

Catalog No. p-7072266
Change view
Click to view available options
Quantity:
100 g
25 g
Catalog No. Quantity
6001521 25 g
6001520 100 g
2 options

Catalog No. 6001521

Supplier: Honeywell Chemicals 7177825G

Only null left

Puriss. p.a., ACS Reagent, Reagent European Pharmacopoeia, Reagent European Pharmacopoeia,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 13755-38-9
Molecular Formula C5H4FeN6Na2O3
Molecular Weight (g/mol) 297.95
MDL Number MFCD00149192
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
PubChem CID 11953895
IUPAC Name disodium;iron(4+);nitroxyl anion;pentacyanide;dihydrate
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

Specifications

Quantity 25 g
Linear Formula Na2[Fe(CN)5NO] · 2H2O
UN Number UN3288
Formula Weight 297.95g/mol
Percent Purity ≥99%
Chemical Name or Material Sodium nitroprusside dihydrate
Hazard Statement P301 + P310
RTECSNumber LJ8925000