missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitroprusside Dihydrate Crystalline MP Biomedicals

Catalog No. ICN15206125
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15206125 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15206125 Supplier MP Biomedicals Inc Supplier No. 0215206125

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Analytical grade chemical reagent for laboratory and research applications.

Chemical Identifiers

CAS 13755-38-9
Molecular Formula C5H4FeN6Na2O3
Molecular Weight (g/mol) 297.95
MDL Number MFCD00149192
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
PubChem CID 11953895
IUPAC Name disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

Specifications

Physical Form Crystalline
Quantity 25 g
Formula Weight 297.95
Percent Purity >99%
Density 7.87 g/cm3 at 25 °C
Chemical Name or Material Sodium Nitroprusside, Dihydrate