missing translation for 'onlineSavingsMsg'
Learn More

Sodium nitroprusside dihydrate, ACS, MilliporeSigma™

Catalog No. M1065410100
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1065410100 100 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1065410100 Supplier Technidata Supplier No. 1.06541.0100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

ACS reagent grade chemical for analytical applications such as spectrophotometric determinations.

Chemical Identifiers

CAS 13755-38-9
Molecular Formula C5H4FeN6Na2O3
Molecular Weight (g/mol) 297.95
MDL Number MFCD00149192
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
PubChem CID 11953895
IUPAC Name disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

Specifications

Color Red
Physical Form Crystals
Quantity 100 g
Linear Formula Na2[Fe(CN)5NO] · 2H2O
Solubility Information 400g/L (20°C)á(slow decomposition)
Formula Weight 297.95
Percent Purity 99.0 to 102.0%
Grade ACS/EP/Reagent
Packaging Plastic Bottle
Density 1.71g/cm3 (20°C); Bulk Density: 1000 kg/m3
Chemical Name or Material Sodium Nitroprusside, For Analysis
Show More Show Less
Hazard Category SignalWord: Danger
Recommended Storage Store without restrictions