missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitroferricyanide, Dihydrate, Crystal, ACS, 99.0- 102.0%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S1365500GM

 View more versions of this product

Catalog No. 18613741


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

13755-38-9
500 g
ACS
1
99.0 to 102.0%
Poly Bottle

Chemical Identifiers

13755-38-9
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.