missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitroferricyanide, Dihydrate, Crystal, ACS, 99.0- 102.0%, Spectrum™ Chemical

Catalog No. 18613739
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
50 kg
Packaging:
Fiber Drum
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613739 25 g Poly Bottle
18613740 125 g Poly Bottle
18613741 500 g Poly Bottle
18613742 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18613739 Supplier Spectrum Chemical Mfg Cor Supplier No. S136525GM
Get Up to $500 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 13755-38-9

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99.0 to 102.0%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.