missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite, Purified, Reagents

Catalog No. p-200020956
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336997 2.5 kg
01336996 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336997 Supplier Labchem Inc Supplier No. C23210002.5A5
Only null left

Sodium Nitrite, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym Nitrous Acid Sodium Salt
IUPAC Name sodium nitrite
SMILES [Na+].[O-]N=O

Specifications

Boiling Point 320°C
Melting Point 270°C
Color White or Slightly Yellow
Physical Form Solid
Quantity 2.5 kg
Grade Pure
Packaging Poly Bottle
Specific Gravity 2.17
Chemical Name or Material Sodium Nitrite, Purified