missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite, Purified, Reagents

Catalog No. 01336996
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
2.5 kg
500 g
Catalog No. Quantity
01336996 500 g
01336997 2.5 kg
2 options

Catalog No. 01336996

Supplier: Labchem Inc C2321000500A5

Only null left

Sodium Nitrite, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym Nitrous Acid Sodium Salt
IUPAC Name sodium nitrite
SMILES [Na+].[O-]N=O

Specifications

Boiling Point 320°C
Melting Point 270°C
Color White or Slightly Yellow
Physical Form Solid
Quantity 500 g
Grade Pure
Packaging Poly Bottle
Specific Gravity 2.17
Chemical Name or Material Sodium Nitrite, Purified