missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1065499012
Change view
Click to view available options
Quantity:
12 kg
Packaging:
Plastic Bucket
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1065499012 12 kg Plastic Bucket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1065499012 Supplier Technidata Supplier No. 1.06549.9012

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Recommended Storage

+5°C to +30°C.

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium nitrite
SMILES [Na+].[O-]N=O

Specifications

Melting Point 280°C (decomposition)
Color White
pH 9 (100g/L, H2O, 20°C)
Physical Form Solid
Quantity 12 kg
Solubility Information 820g/L
Formula Weight 69
Percent Purity >99.0%
Packaging Plastic Bucket
Density 2.1 g/3 (20 °C)
Chemical Name or Material Sodium Nitrite
Show More Show Less
EINECSNumber 007-010-00-4
RTECSNumber RA1225000
Recommended Storage +5°C to +30°C.