missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. p-34028
Change view
Click to view available options
Quantity:
250 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S347250 250 g Poly Bottle
S347500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S347250 Supplier Thermo Fisher Scientific Supplier No. S347250
Only null left

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium nitrite
SMILES [Na+].[O-]N=O

Specifications

Boiling Point 320°C
Melting Point 271°C
Color Yellow
pH 8 to 9
Physical Form Solid
Quantity 250 g
Assay Percent Range ≥97 %
Formula Weight 69
Percent Purity ≥97%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Decomposition Information >320°C
Chemical Name or Material Sodium Nitrite
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Oxidizer: Contact with combustible/organic material may cause fire. May cause central nervous system effects. May cause cyanosis. May cause methemoglobinemia. May cause skin, eye, and respiratory tract irritation. Very toxic to aquatic organisms. Use personal protective equipment. Ensure adequate ventilation. Keep away from clothing and other combustible materials. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention.

NFPA

Health:3
Flammability:0
Instability:2
DOTInformation DOT Class 5.1, : Oxidizer