missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite, Crystal, ACS, 97%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor S1355125GM

 View more versions of this product

Catalog No. 18613730


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

7632-00-0
125 g
ACS
1
97%
Poly Bottle

Chemical Identifiers

7632-00-0
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.