missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite, Crystal, ACS, 97%, Spectrum™ Chemical

Catalog No. 18613729
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613729 25 g Poly Bottle
18613730 125 g Poly Bottle
18613731 500 g Poly Bottle
18613732 2.5 kg Poly Bottle
18613733 12 kg Poly Pail
18613734 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18613729 Supplier Spectrum Chemical Mfg Cor Supplier No. S135525GM

Chemical Identifiers

CAS 7632-00-0

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 97%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.