missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrite, ACS Reagent, Reagents

Catalog No. 01337708
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337708 2.5 kg
01337707 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337708 Supplier Labchem Inc Supplier No. C13210002.5A5

Sodium Nitrite, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym Nitrous Acid Sodium Salt
IUPAC Name sodium nitrite
SMILES [Na+].[O-]N=O

Specifications

Boiling Point 320°C
Melting Point 270°C
Color White or Slightly Yellow
Physical Form Solid
Quantity 2.5 kg
Grade ACS
Packaging Poly Bottle
Specific Gravity 2.17
Chemical Name or Material Sodium Nitrite, ACS Reagent