Learn More
Sodium nitrite, 98%
CAS: 7632-00-0 | NaNO2 | 69.00 g/mol
Supplier: Thermo Scientific Chemicals A186680B
Description
Sodium nitrite is used in many industrial applications viz. in meat curing, coloring and preserving. It is used as a reagent in analytical chemistry, an antidote in cyanide poisoning, an electrolyte in electrochemical grinding, as a cooling solution in closed loop systems and as an additive in industrial greases. It finds application as a corrosion inhibitor as well as in the rubber industry. In metallurgy, it is used for phosphatizing and detinning. It acts as a precursor to diazo dyes, nitroso compounds and various organic compounds like pharmaceuticals. As a food additive, it is used to prevent botulism.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium nitrite is used in many industrial applications viz. in meat curing, coloring and preserving. It is used as a reagent in analytical chemistry, an antidote in cyanide poisoning, an electrolyte in electrochemical grinding, as a cooling solution in closed loop systems and as an additive in industrial greases. It finds application as a corrosion inhibitor as well as in the rubber industry. In metallurgy, it is used for phosphatizing and detinning. It acts as a precursor to diazo dyes, nitroso compounds and various organic compounds like pharmaceuticals. As a food additive, it is used to prevent botulism.
Solubility
Soluble in water, methanol, ethanol, ammonia, nitric acid. Slightly soluble diethyl ether.
Notes
Decomposes above 320°. Hygroscopic. Incompatible with reducing agents, strong oxidizing agents, organics, finely powdered metals.
Specifications
| Sodium nitrite | |
| 7632-00-0 | |
| NaNO2 | |
| MFCD00011118 | |
| 14,8648 | |
| Soluble in water,methanol,ethanol,ammonia,nitric acid. Slightly soluble diethyl ether. | |
| [Na+].[O-]N=O | |
| 69.00 | |
| CHEBI:78870 | |
| 98% | |
| Hygroscopic |
| 271°C (decomposition) | |
| 1000 g | |
| NaNO2 | |
| UN1500 | |
| sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts | |
| LPXPTNMVRIOKMN-UHFFFAOYSA-M | |
| sodium nitrite | |
| 23668193 | |
| 69 | |
| Odorless | |
| 2.17 |
Chemical Identifiers
| 7632-00-0 | |
| 69.00 | |
| LPXPTNMVRIOKMN-UHFFFAOYSA-M | |
| 23668193 | |
| sodium nitrite |
| NaNO2 | |
| MFCD00011118 | |
| sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts | |
| CHEBI:78870 | |
| [Na+].[O-]N=O |
Safety and Handling
GHS H Statement
H301-H272
Toxic if swallowed.
May intensify fire; oxidizer.
P210-P220-P221-P260-P264b-P270-P271-P280i-P281-P301+P310-P304+P340-P305+P351+P338-P308+P313-P330-P370+P378q-P501c
H272-H301-H319-H373
DOTInformation : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: SODIUM NITRITE
EINECSNumber : 231-555-9
RTECSNumber : RA1225000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only