missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nicotinate 98.0+%, TCI America™

Catalog No. N0089500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N0089500G 500 g
N008925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N0089500G Supplier TCI America Supplier No. N0089500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 54-86-4 | C6H4NNaO2 | 145.093 g/mol

Chemical Identifiers

CAS 54-86-4
Molecular Formula C6H4NNaO2
Molecular Weight (g/mol) 145.093
MDL Number MFCD00064352
InChI Key KFLRWGSAMLBHBV-UHFFFAOYSA-M
Synonym sodium nicotinate, nicotinic acid sodium salt, sodium pyridine-3-carboxylate, natriumnicotinat german, unii-3zx78b4q2l, natriumnicotinat, 3-pyridinecarboxylic acid, sodium salt, nicotinic acid, sodium salt, 3-pyridinecarboxylic acid, sodium salt 1:1, nicotinic acid sodium
PubChem CID 23687810
IUPAC Name sodium;pyridine-3-carboxylate
SMILES C1=CC(=CN=C1)C(=O)[O-].[Na+]

Specifications

Color White
Quantity 500 g
Formula Weight 145.09
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Nicotinate
EINECSNumber (5)-6624
RTECSNumber QT2060000
TSCA Yes