Learn More
Sodium n-tridecyl sulfate, 99%, may cont. up to ca 5% water and residual solvent
CAS: 3026-63-9 | C13H27NaO4S | 302.405 g/mol
Supplier: Thermo Scientific Chemicals B2208906
Description
Sodium n-tridecyl sulfate acts as a foaming agent and a wetting agent. It is used as a cleaning agent and finds application in industrial and household cleaning products and personal care products. Further, it serves as an emulsifying agent in the transport of hydrocarbons. In addition to this, it acts as an anionic surfactant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium n-tridecyl sulfate acts as a foaming agent and a wetting agent. It is used as a cleaning agent and finds application in industrial and household cleaning products and personal care products. Further, it serves as an emulsifying agent in the transport of hydrocarbons. In addition to this, it acts as an anionic surfactant.
Solubility
Soluble in water.
Notes
Incompatible with bases and strong oxidizing agents.
Specifications
| Sodium n-tridecyl sulfate | |
| 185°C to 189°C | |
| C13H27NaO4S | |
| MFCD00024993 | |
| sodium tridecyl sulfate, unii-jt30s80yr4, sodium n-tridecyl sulphate, sulfuric acid, mono-c8-18-alkyl esters, sodium salts, 1-tridecanol, 1-hydrogen sulfate , sodium salt 1:1, tridecyl sodium sulfate, sodium tridecyl sulphate, 1-tridecanol, hydrogen sulfate, sodium salt, sodium mono-c8-18-alkyl sulfates, sodium tridecylsulfate | |
| HQCFDOOSGDZRII-UHFFFAOYSA-M | |
| sodium;tridecyl sulfate | |
| 23668353 | |
| 99% |
| 3026-63-9 | |
| may cont. up to ≈2% water and residual solvent | |
| CH3(CH2)12OSO3Na | |
| 5 g | |
| Soluble in water. | |
| CCCCCCCCCCCCCOS(=O)(=O)[O-].[Na+] | |
| 302.405 | |
| 302.4 |
Chemical Identifiers
| 3026-63-9 | |
| 302.405 | |
| HQCFDOOSGDZRII-UHFFFAOYSA-M | |
| 23668353 | |
| CCCCCCCCCCCCCOS(=O)(=O)[O-].[Na+] |
| C13H27NaO4S | |
| MFCD00024993 | |
| sodium tridecyl sulfate, unii-jt30s80yr4, sodium n-tridecyl sulphate, sulfuric acid, mono-c8-18-alkyl esters, sodium salts, 1-tridecanol, 1-hydrogen sulfate , sodium salt 1:1, tridecyl sodium sulfate, sodium tridecyl sulphate, 1-tridecanol, hydrogen sulfate, sodium salt, sodium mono-c8-18-alkyl sulfates, sodium tridecylsulfate | |
| sodium;tridecyl sulfate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 221-188-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only