Learn More
Sodium n-octyl sulfate, 99%
CAS: 142-31-4 | C8H17NaO4S | 232.27 g/mol
Supplier: Thermo Scientific Chemicals A1178606
Description
Sodium n-octyl sulfate is a useful chemical to study the selective enrichment of albumin in biological samples and to determine the mixed solution of a glycerin-modified cationic surfactant and an anionic surfactant. It is also used as wetting agent, detergent, dispersant, emulsifier, degreasing agent, hard surface cleaner, lime soap dispersant, alkaline detergent, cleaning agent for metal processing in electroplating, latex, metal processing, paper, polymer synthesis, textile, leather and other industries. It finds application as an anionic detergent and ion -pairing reagent used in high-performance liquid chromatography (HPLC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium n-octyl sulfate is a useful chemical to study the selective enrichment of albumin in biological samples and to determine the mixed solution of a glycerin-modified cationic surfactant and an anionic surfactant. It is also used as wetting agent, detergent, dispersant, emulsifier, degreasing agent, hard surface cleaner, lime soap dispersant, alkaline detergent, cleaning agent for metal processing in electroplating, latex, metal processing, paper, polymer synthesis, textile, leather and other industries. It finds application as an anionic detergent and ion -pairing reagent used in high-performance liquid chromatography (HPLC).
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Sodium n-octyl sulfate | |
| ∼195°C (decomposition) | |
| MFCD00007470 | |
| 3658635 | |
| Soluble in water. | |
| CCCCCCCCOS(=O)(=O)[O-].[Na+] | |
| 232.27 | |
| 232.28 |
| 142-31-4 | |
| C8H17NaO4S | |
| 5 g | |
| sodium octyl sulfate, sodium n-octyl sulfate, sodium octyl sulphate, sipex ols, cycloryl os, duponol 80, sodium capryl sulfate, octyl sodium sulfate, sulfuric acid, monooctyl ester, sodium salt, octyl sulfate sodium salt | |
| WFRKJMRGXGWHBM-UHFFFAOYSA-M | |
| sodium;octyl sulfate | |
| 2735107 | |
| 99% |
Chemical Identifiers
| 142-31-4 | |
| 232.27 | |
| WFRKJMRGXGWHBM-UHFFFAOYSA-M | |
| 2735107 | |
| CCCCCCCCOS(=O)(=O)[O-].[Na+] |
| C8H17NaO4S | |
| MFCD00007470 | |
| sodium octyl sulfate, sodium n-octyl sulfate, sodium octyl sulphate, sipex ols, cycloryl os, duponol 80, sodium capryl sulfate, octyl sodium sulfate, sulfuric acid, monooctyl ester, sodium salt, octyl sulfate sodium salt | |
| sodium;octyl sulfate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
H315-H318
EINECSNumber : 205-535-5
RTECSNumber : WT1175000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only