Learn More
Sodium n-decyl sulfate, 98%
CAS: 142-87-0 | C10H21NaO4S | 260.324 g/mol
Supplier: Thermo Scientific Chemicals A1629914
Description
sodium decyl sulphate is used as a superior dispersing, wetting and emulsifying agent suitable for use in the production of gypsum boards, fire-fighting foams, carpet and upholstery cleaners, special industrial and all-purpose cleaners, oil fields and in the latex manufacture. It is also been used in it to be used in ion pair chromatography and icellar electrokinetic capillary chromatography. other applications include HI&I cleaning, emulsion polymerization, Fire-fighting, Industrial auxiliaries, Construction chemicals, Oil fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationssodium decyl sulphate is used as a superior dispersing, wetting and emulsifying agent suitable for use in the production of gypsum boards, fire-fighting foams, carpet and upholstery cleaners, special industrial and all-purpose cleaners, oil fields and in the latex manufacture. It is also been used in it to be used in ion pair chromatography and icellar electrokinetic capillary chromatography. other applications include HI&I cleaning, emulsion polymerization, Fire-fighting, Industrial auxiliaries, Construction chemicals, Oil fields.
Solubility
It is soluble in H20, poorly soluble in organic solvents.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Hygroscopic, Keepp away from acids Strong oxidizing agents.
Specifications
| Sodium n-decyl sulfate | |
| C10H21NaO4S | |
| 25 g | |
| sodium decyl sulfate, sodium n-decyl sulfate, decyl sodium sulfate, sulfuric acid, monodecyl ester, sodium salt, sodium decyl sulphate, unii-al92m833sy, n-decyl sodium sulfate, sodium n-decylsulphate, sulfuric acid, monodecyl ester, sodium salt 1:1, sulfuric acid, decyl ester, sodium salt | |
| XZTJQQLJJCXOLP-UHFFFAOYSA-M | |
| sodium;decyl sulfate | |
| 23665771 | |
| 99% |
| 142-87-0 | |
| MFCD00041881 | |
| 3576860 | |
| It is soluble in H20,poorly soluble in organic solvents. | |
| CCCCCCCCCCOS(=O)(=O)[O-].[Na+] | |
| 260.324 | |
| 260.33 |
Chemical Identifiers
| 142-87-0 | |
| 260.324 | |
| XZTJQQLJJCXOLP-UHFFFAOYSA-M | |
| 23665771 | |
| CCCCCCCCCCOS(=O)(=O)[O-].[Na+] |
| C10H21NaO4S | |
| MFCD00041881 | |
| sodium decyl sulfate, sodium n-decyl sulfate, decyl sodium sulfate, sulfuric acid, monodecyl ester, sodium salt, sodium decyl sulphate, unii-al92m833sy, n-decyl sodium sulfate, sodium n-decylsulphate, sulfuric acid, monodecyl ester, sodium salt 1:1, sulfuric acid, decyl ester, sodium salt | |
| sodium;decyl sulfate |
Safety and Handling
GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P501c
H302-H315-H318-H335
EINECSNumber : 205-568-5
RTECSNumber : WS7625000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only