missing translation for 'onlineSavingsMsg'
Learn More

Sodium Monododecyl Phosphate (mixture of Mono and Disodium Salt), TCI America™

Catalog No. P086525G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P086525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P086525G Supplier TCI America Supplier No. P086525G
Only null left

Chemical Identifiers

CAS 7423-32-7
Molecular Formula C12H25Na2O4P
Molecular Weight (g/mol) 310.281
MDL Number MFCD00058971
InChI Key YVIGPQSYEAOLAD-UHFFFAOYSA-L
Synonym Phosphoric Acid Monododecyl Ester Sodium Salt, Sodium Monolauryl Phosphate
PubChem CID 6452307
IUPAC Name disodium;dodecyl phosphate
SMILES CCCCCCCCCCCCOP(=O)([O-])[O-].[Na+].[Na+]

Specifications

Color White
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Sodium Monododecyl Phosphate (mixture of Mono and Disodium Salt)