missing translation for 'onlineSavingsMsg'
Learn More

Sodium molybdenum oxide dihydrate, ACS, 99.5-103.0%

Catalog No. p-4407917
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1221422 100 g
AA1221436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1221422 Supplier Thermo Scientific Chemicals Supplier No. 01221422
Only null left

CAS: 10102-40-6 | MoNa2O4 | 205.93 g/mol

It is widely used as a fertilizer in agriculture, and a corrosion inhibitor being a non-oxidizing anodic inhibitor. It reduces galvanic corrosion potential, useful in industrial waste water treatment. It is recommended for treatment of whiptail in broccoli and cauliflower. It is a precursor for molybdenum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10102-40-6
Molecular Formula H4MoNa2O6
Molecular Weight (g/mol) 241.96
MDL Number MFCD00149170
InChI Key FDEIWTXVNPKYDL-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
PubChem CID 16211258
ChEBI CHEBI:75213
IUPAC Name disodium dioxomolybdenumbis(olate) dihydrate
SMILES O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Melting Point 100°C-2H2O
Color Green
Physical Form Powder
Quantity 100 g
Assay Percent Range 99.5-103.0%
Solubility Information Soluble in water.
Formula Weight 241.95 (205.92 Anhydrous)
Grade ACS
Odor Odorless
Appearance White powder/flake/crystalline/beads
Density 2.71
Chemical Name or Material Sodium molybdenum oxide dihydrate, For analysis ACS
Show More Show Less
EINECSNumber 231-551-7
RTECSNumber QA5085000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only