missing translation for 'onlineSavingsMsg'
Learn More

Sodium molybdenum oxide dihydrate, 98%

Catalog No. p-7024482
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1922222 100 g
AAA1922236 500 g
AAA192220E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1922222 Supplier Thermo Scientific Chemicals Supplier No. A1922222
Only null left

CAS: 10102-40-6 | MoNa2O4 | 205.93 g/mol

Sodium molybdenum oxide is widely used as a fertilizer in agriculture, and a corrosion inhibitor being a non-oxidizing anodic inhibitor. It reduces galvanic corrosion potential, useful in industrial waste water treatment. It is recommended for treatment of whiptail in broccoli and cauliflower. It is a precursor for molybdenum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium molybdenum oxide is widely used as a fertilizer in agriculture, and a corrosion inhibitor being a non-oxidizing anodic inhibitor. It reduces galvanic corrosion potential, useful in industrial waste water treatment. It is recommended for treatment of whiptail in broccoli and cauliflower. It is a precursor for molybdenum.

Solubility
Soluble in water.

Notes
Stable. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 10102-40-6
Molecular Formula H4MoNa2O6
Molecular Weight (g/mol) 241.96
MDL Number MFCD00149170
InChI Key FDEIWTXVNPKYDL-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
PubChem CID 16211258
ChEBI CHEBI:75213
IUPAC Name disodium dioxomolybdenumbis(olate) dihydrate
SMILES O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Melting Point 100°C (in -2H2O)
Quantity 100 g
Merck Index 14,8645
Solubility Information Soluble in water.
Formula Weight 241.95 (205.92 Anhydrous)
Percent Purity 98%
Odor Odorless
Density 2.71
Chemical Name or Material Sodium molybdenum oxide dihydrate
EINECSNumber 231-551-7
RTECSNumber QA5085000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only