missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium molybdate(VI) dihydrate, ACS reagent
CAS: 10102-40-6 | MoNa2O4 | 205.93 g/mol
$90.12 - $475.78
Chemical Identifiers
| CAS | 10102-40-6 |
|---|---|
| Molecular Formula | H4MoNa2O6 |
| Molecular Weight (g/mol) | 241.96 |
| MDL Number | MFCD00149170 |
| InChI Key | FDEIWTXVNPKYDL-UHFFFAOYSA-N |
| Synonym | sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate |
| PubChem CID | 16211258 |
| ChEBI | CHEBI:75213 |
| IUPAC Name | disodium dioxomolybdenumbis(olate) dihydrate |
| SMILES | O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC446360250
|
Thermo Scientific Chemicals
446360250 |
25 g | Plastic bottle |
Each for $90.12
|
|
||||
|
AC446361000
|
Thermo Scientific Chemicals
446361000 |
100 g | Plastic bottle |
Each for $204.13
|
|
||||
|
AC446365000
|
Thermo Scientific Chemicals
446365000 |
500 g | Plastic bottle |
Each for $475.78
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 10102-40-6 | |
| 241.96 | |
| FDEIWTXVNPKYDL-UHFFFAOYSA-N | |
| 16211258 | |
| disodium dioxomolybdenumbis(olate) dihydrate |
| H4MoNa2O6 | |
| MFCD00149170 | |
| sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate | |
| CHEBI:75213 | |
| O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O |
Specifications
| 10102-40-6 | |
| H4MoNa2O6 | |
| MFCD00149170 | |
| sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate | |
| FDEIWTXVNPKYDL-UHFFFAOYSA-N | |
| disodium dioxomolybdenumbis(olate) dihydrate | |
| 16211258 | |
| 241.95 | |
| Plastic bottle | |
| 3.2800g/mL |
| 25 g | |
| Na2MoO4·2H2O | |
| 15,8776 | |
| Solubility in water: 560g/L (o°C). Other solubilities: 1150g/l (100°C) | |
| O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O | |
| 241.96 | |
| CHEBI:75213 | |
| ACS Reagent | |
| 3.28 | |
| Sodium molybdate(VI) dihydrate |
RUO – Research Use Only