missing translation for 'onlineSavingsMsg'
Learn More

Sodium molybdate(VI) dihydrate, 99+%

Catalog No. p-221981
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC206370050 5 g Glass Bottle
AC206371000 100 g Plastic bottle
AC206375000 500 g Plastic bottle
AC206370025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC206370050 Supplier Thermo Scientific Chemicals Supplier No. 206370050

CAS: 10102-40-6 | MoNa2O4 | 205.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent

Chemical Identifiers

CAS 10102-40-6
Molecular Formula H4MoNa2O6
Molecular Weight (g/mol) 241.96
MDL Number MFCD00149170
InChI Key FDEIWTXVNPKYDL-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
PubChem CID 16211258
ChEBI CHEBI:75213
IUPAC Name disodium dioxomolybdenumbis(olate) dihydrate
SMILES O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Color White
Physical Form Crystalline Powder
Quantity 5 g
Assay Percent Range 99% min
Linear Formula Na2MoO4·2H2O
Fieser 15,294
Merck Index 15, 8776
Solubility Information Solubility in water: 560g/L (o°C). Other solubilities: 1150g/l (100°C)
Formula Weight 241.95
Percent Purity ≥99%
Packaging Glass Bottle
Specific Gravity 3.28
Density 3.2800g/mL
Chemical Name or Material Sodium molybdate(VI) dihydrate, 99+%
Show More Show Less
RTECSNumber QA5085000

RUO – Research Use Only