missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Molybdate Dihydrate, puriss., Solstice
Description
- Leading the Industry For Over 65 Years
- Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Physical Form | Crystalline |
| Quantity | 1 kg |
| Linear Formula | Na2MoO4 · 2H2O |
| UN Number | NONH for all modes of transport |
| Formula Weight | 241.95g/mol |
| Percent Purity | 99 to 100.5% (calc. to the dried substance) |
| Grade | puriss. |
| Chemical Name or Material | Sodium Molybdate Dihydrate, Meets analytical specification of PhEur |
Safety and Handling
| EINECSNumber | 231-551-7 |