missing translation for 'onlineSavingsMsg'
Learn More

Sodium Molybdate Dihydrate (Crystalline Powder/Certified ACS), Fisher Chemical™

Catalog No. p-33972
Change view
Click to view available options
Quantity:
500 g
3 kg
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S336500 500 g Glass Bottle
S3363 3 kg Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S336500 Supplier Thermo Fisher Scientific Supplier No. S336 500
Only null left

Chemical Identifiers

CAS 10102-40-6
Molecular Formula H4MoNa2O6
Molecular Weight (g/mol) 241.96
MDL Number MFCD00149170
InChI Key FDEIWTXVNPKYDL-UHFFFAOYSA-N
Synonym sodium molybdate dihydrate, sodium molybdate vi dihydrate, disodium molybdate dihydrate, unii-8f2sxi1704, na2moo4.2h2o, disodium dihydrate molybdate, molybdic acid sodium salt dihydrate, molybdic acid, disodium salt, dihydrate, sodium molybdate 2 hydrate, dipotassium dihydrate molybdate
PubChem CID 16211258
ChEBI CHEBI:75213
IUPAC Name disodium dioxomolybdenumbis(olate) dihydrate
SMILES O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Color White
Physical Form Solid
Quantity 500 g
Assay Percent Range 99.5 to 103.0%
Formula Weight 241.95
Percent Purity 99.5 to 103.0%
Grade Certified ACS
Identification Pass Test
Packaging Glass Bottle
Density 3.28g/cm³
Chemical Name or Material Sodium Molybdate Dihydrate
Show More Show Less