missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Molybdate Dihydrate (Crystalline Powder/Certified ACS), Fisher Chemical™
Specifications
Specifications
| Color | White |
| Physical Form | Solid |
| Quantity | 500 g |
| Identification | Pass Test |
| Packaging | Glass Bottle |
| Assay Percent Range | 99.5 to 103.0% |
| Density | 3.28g/cm³ |
| Formula Weight | 241.95 |
| Percent Purity | 99.5 to 103.0% |
| Grade | Certified ACS |
| Show More |