missing translation for 'onlineSavingsMsg'
Learn More

Sodium Molybdate, Dihydrate, Crystal, CP, 98%, Spectrum™ Chemical

Catalog No. 18613716
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
200 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613716 2.5 kg Poly Bottle
18613714 125 g Poly Bottle
18613715 500 g Poly Bottle
18613719 200 lb. Fiber Drum
18613717 12 kg Poly Pail
18613718 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18613716 Supplier Spectrum Chemical Mfg Cor Supplier No. S13422.5KG
Only null left

Chemical Identifiers

CAS 10102-40-6

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 98%
Grade Chemically Pure
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.