missing translation for 'onlineSavingsMsg'
Learn More

Sodium Molybdate, Dihydrate, Crystal, CP, 98%, Spectrum™ Chemical

Catalog No. 18613714
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
200 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18613714 125 g Poly Bottle
18613715 500 g Poly Bottle
18613719 200 lb. Fiber Drum
18613716 2.5 kg Poly Bottle
18613717 12 kg Poly Pail
18613718 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18613714 Supplier Spectrum Chemical Mfg Cor Supplier No. S1342125GM

Chemical Identifiers

CAS 10102-40-6

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 98%
Grade Chemically Pure
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.