missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Molybdate, Dihydrate, Crystal, ACS, 99.5-103.0%, Spectrum™ Chemical
Specifications
Specifications
| pH | 7.0 to 10.5 |
| CAS Max % | 1 |
| Quantity | 12 kg |
| Percent Purity | 99.5 to 103.0% |
| Grade | ACS |
| Packaging | Poly Pail |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.