missing translation for 'onlineSavingsMsg'
Learn More

Sodium Molybdate Dihydrate, ACS Reagent, Reagents

Catalog No. 01337713
Change view
Click to view available options
Quantity:
100 g
500 g
Catalog No. Quantity
01337713 500 g
01337714 100 g
2 options

Catalog No. 01337713

Supplier: Labchem Inc C1320300500C2

Only null left

Sodium Molybdate Dihydrate, ACS Reagent, ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 10102-40-6
Molecular Formula H4MoNa2O6
Molecular Weight (g/mol) 241.96
MDL Number MFCD00149170
InChI Key FDEIWTXVNPKYDL-UHFFFAOYSA-N
IUPAC Name disodium dioxomolybdenumbis(olate) dihydrate
SMILES O.O.[Na+].[Na+].[O-][Mo]([O-])(=O)=O

Specifications

Boiling Point >687°C
Color White
Physical Form Solid
Quantity 500 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 3.5
Chemical Name or Material Sodium Molybdate Dihydrate, ACS Reagent