missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Methoxide Solution, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 124-41-4 |
| Quantity | 100 mL |
| Molecular Formula | CH3NaO |
| Linear Formula | CH3ONa |
| MDL Number | MFCD00012179 |
| UN Number | UN1289 - Class 3 - PG 2 - Sodium Methylate Solutions |
| Flash Point | 9.7°C (Closed Cup) |
| Synonym | Methanolic Base; Sodium Methylate |
| Grade | GC Derivatization |
| Concentration or Composition (by Analyte or Components) | 0.5 M |
| Show More |