missing translation for 'onlineSavingsMsg'
Learn More

Sodium Methanesulfonate 98.0+%, TCI America™

Catalog No. S051825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S051825G 25 g
S0518500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S051825G Supplier TCI America Supplier No. S051825G

Chemical Identifiers

CAS 2386-57-4
Molecular Formula CH3NaO3S
Molecular Weight (g/mol) 118.082
InChI Key KKVTYAVXTDIPAP-UHFFFAOYSA-M
Synonym sodium methanesulfonate, methanesulfonic acid sodium salt, methanesulfonic acid, sodium salt, sodium methanesulphonate, sodium mesylate, methylsulfonic acid sodium salt, 75-75-2 parent, sodium methyl sulfonate, methanesulfonic acid, sodium salt 1:1, sodium methylsulfonate
PubChem CID 638112
IUPAC Name sodium;methanesulfonate
SMILES CS(=O)(=O)[O-].[Na+]

Specifications

Color White
Quantity 25 g
Formula Weight 118.08
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Methanesulfonate
EINECSNumber (2)-4084&(2)-4141
TSCA Yes