missing translation for 'onlineSavingsMsg'
Learn More

Sodium methanesulfonate, 99%

Catalog No. AC442110050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC442110050 5 g
AC442111000 100 g
AC442115000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC442110050 Supplier Thermo Scientific Chemicals Supplier No. 442110050

CAS: 2386-57-4 | CH4NaO3S | 118.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2386-57-4
Molecular Formula CH4NaO3S
Molecular Weight (g/mol) 118.09
InChI Key KKVTYAVXTDIPAP-UHFFFAOYSA-M
Synonym sodium methanesulfonate, methanesulfonic acid sodium salt, methanesulfonic acid, sodium salt, sodium methanesulphonate, sodium mesylate, methylsulfonic acid sodium salt, 75-75-2 parent, sodium methyl sulfonate, methanesulfonic acid, sodium salt 1:1, sodium methylsulfonate
PubChem CID 638112
IUPAC Name sodium;methanesulfonate
SMILES CS(=O)(=O)[O-].[Na+]

Specifications

Quantity 5 g
Linear Formula CH3SO3Na
Solubility Information Solubility in water: soluble.
Formula Weight 118.09
Percent Purity 99%
Infrared Spectrum Authentic
Chemical Name or Material Sodium methanesulfonate
Hazard Category Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
EINECSNumber 219-203-2

RUO – Research Use Only