missing translation for 'onlineSavingsMsg'
Learn More

Sodium methanesulfonate, 98%

Catalog No. AAB2243914
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2243914 25 g
AAB2243906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2243914 Supplier Thermo Scientific Chemicals Supplier No. B2243914

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2386-57-4 | CH3NaO3S | 118.082 g/mol

Sodium methanesulfonate is used as a non-steroidal anti-inflammatory drug.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium methanesulfonate is used as a non-steroidal anti-inflammatory drug.

Solubility
Soluble in water, alcohol, ether, hexane, benzene, methylcyclopentane, toluene, o-chlorotoluene and ethyl disulfide.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 2386-57-4
Molecular Formula CH3NaO3S
Molecular Weight (g/mol) 118.082
MDL Number MFCD00064389
InChI Key KKVTYAVXTDIPAP-UHFFFAOYSA-M
Synonym sodium methanesulfonate, methanesulfonic acid sodium salt, methanesulfonic acid, sodium salt, sodium methanesulphonate, sodium mesylate, methylsulfonic acid sodium salt, 75-75-2 parent, sodium methyl sulfonate, methanesulfonic acid, sodium salt 1:1, sodium methylsulfonate
PubChem CID 638112
IUPAC Name sodium;methanesulfonate
SMILES CS(=O)(=O)[O-].[Na+]

Specifications

Odor Odorless
Linear Formula CH3SO3Na
Quantity 25 g
Sensitivity Hygroscopic
Solubility Information Soluble in water,alcohol,ether,hexane,benzene,methylcyclopentane,toluene,o-chlorotoluene and ethyl disulfide.
Formula Weight 118.09
Percent Purity 98%
Chemical Name or Material Sodium methanesulfonate
EINECSNumber 219-203-2
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only