missing translation for 'onlineSavingsMsg'
Learn More

Sodium Methanesulfinate 90.0+%, TCI America™

Catalog No. p-7132623
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12555G 5 g
M125525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M12555G Supplier TCI America Supplier No. M12555G
Only null left

Chemical Identifiers

CAS 20277-69-4
Molecular Formula CH3NaO2S
Molecular Weight (g/mol) 102.083
MDL Number MFCD00040392
InChI Key LYPGDCWPTHTUDO-UHFFFAOYSA-M
Synonym sodium methanesulfinate, sodium methanesulphinate, methanesulfinic acid sodium salt, sodium methylsulfinate, methanesulfinic acid, sodium salt, sodium methane sulfinate, methanesulfinicacidsodiumsalt, methanesulphinic acid sodium salt, methane sulfinic acid sodium salt, meso2na
PubChem CID 2733271
IUPAC Name sodium;methanesulfinate
SMILES CS(=O)[O-].[Na+]

Specifications

Melting Point 226°C
Color White-Yellow
Quantity 5 g
Formula Weight 102.08
Percent Purity ≥90.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Methanesulfinate