missing translation for 'onlineSavingsMsg'
Learn More

Sodium metavanadate, typically 96%, V 38% min

CAS: 13718-26-8 | NaO3V | 121.93 g/mol

Supplier:  Thermo Scientific Chemicals 01109230

 View more versions of this product

Catalog No. AA1109230


Only null left
Explore available promotions

Description

Description

Sodium metavanadate is used as an analytical reagent and mordant. It is also found to be useful in the photography industry and the pharmaceutical industry. In addition to this, it is used in the making of vanadium alloy and vanadium catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium metavanadate
V 38% min.
White
250 g
NaO3V
sodium metavanadate, sodium vanadate v, vanadic acid, monosodium salt, sodium vanadate meta, sodium oxido dioxo vanadium, sodium trioxovanadate 1-, metawanadan sodowy polish, vanadate vo31-, sodium, monosodium trioxovanadate 1-, ccris 6881
CMZUMMUJMWNLFH-UHFFFAOYSA-N
sodium vanadiumoylolate
4148882
121.93
Off-white to light orange powder
630°C
13718-26-8
Chunks and/or Powder
Typically 96%
MFCD00011125
Soluble in water.
[Na+].[O-][V](=O)=O
121.93
CHEBI:75221
Odorless
2.79g/cm3 at 20°C

Chemical Identifiers

13718-26-8
121.93
CMZUMMUJMWNLFH-UHFFFAOYSA-N
4148882
[Na+].[O-][V](=O)=O
NaO3V
MFCD00011125
sodium metavanadate, sodium vanadate v, vanadic acid, monosodium salt, sodium vanadate meta, sodium oxido dioxo vanadium, sodium trioxovanadate 1-, metawanadan sodowy polish, vanadate vo31-, sodium, monosodium trioxovanadate 1-, ccris 6881
CHEBI:75221
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P310-P304+P340-P305+P351+P338-P308+P313-P330-P501c

H301-H319-H332-H361-H372-H500

DOTInformation : DOT Class: 6.1; Packaging Group: II

EINECSNumber : 237-272-7

RTECSNumber : YW1050000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only