missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium metatungstate hydrate, crystalline
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White to Yellow |
| Physical Form | Crystalline Powder or Beads |
| Quantity | 50 g |
| Linear Formula | Na6[H2W12O40]·xH2O |
| Solubility Information | Solubility in water: >1000g/L (20°C). |
| Formula Weight | 2986.13 |
| Percent Purity | ≥85% (WO3) |
| Packaging | Glass bottle |
| Chemical Name or Material | Sodium metatungstate hydrate |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Harmful if swallowed. Causes serious eye damage. Harmful to aquatic life with long lasting effects. |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid release to the environment. |
RUO – Research Use Only