missing translation for 'onlineSavingsMsg'
Learn More

Sodium metatungstate, 2.82 density solution in water, stabilized, Thermo Scientific™

Chemical Identifiers

CAS 12141-67-2
Molecular Formula H2Na6O40W12
MDL Number MFCD00150663
Synonym Sodium polytungstate
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC304100500
SDS View Documents
Thermo Scientific Chemicals
304100500
50mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC304102500
SDS View Documents
Thermo Scientific Chemicals
304102500
250mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

12141-67-2
H2Na6O40W12
MFCD00150663
Sodium polytungstate

Specifications

12141-67-2
Na6[H2W12O40]
2.8200g/mL
2986.13
Glass bottle
50mL
H2Na6O40W12
MFCD00150663
Sodium polytungstate
≥67% (WO3)
2.82
Sodium metatungstate, 2.82 Density Solution in Water, Stabilized
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Harmful to aquatic life with long lasting effects.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only