missing translation for 'onlineSavingsMsg'
Learn More

Sigma Aldrich Sodium Metatitanate

Catalog No. 40130750G
Encompass
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40130750G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40130750G Supplier Sigma Aldrich Supplier No. 40130750G

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used as a low-voltage sodium titanium oxide insertion electrode for sodium-ion batteriesUsed for preparation of:Titania nanorods and nanowires via a hydrothermal method for use in lithium-ion storage batteriesPhotocatalytic titanate nanotube thin films via a hydrothermal methodProtonated titanate nanotubes as solid acid catalystsNano-sized supports of ruthenium nanoparticles for catalytic ammonia decomposition

Specifications

CAS 13314-52-8
Density 1.372 g/mL at 25°C
Molecular Formula C4H10Cl2Ge
Linear Formula (C2H5)2GeCl2
Quantity 50 g
MDL Number MFCD00013589
Synonym Sodium titanium oxide
Molecular Weight (g/mol) 201.67 g/mol
Biological Activity Respiratory System