missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Metasilicate Pentahydrate, Purified, Reagents
Specifications
Specifications
| Boiling Point | >1200°C |
| Color | Colorless/White |
| Physical Form | Solid |
| Quantity | 3 kg |
| Grade | Reagent |
| Packaging | Poly Bottle |
| Specific Gravity | 1.52 |
| Chemical Name or Material | Sodium Metasilicate Pentahydrate, Purified |