missing translation for 'onlineSavingsMsg'
Learn More

Sodium metasilicate nonahydrate, 44-47.5% total solids

Catalog No. AC419590250
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC419590250 25 g Plastic bottle
AC419595000 500 g Plastic bottle
AC419590025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC419590250 Supplier Thermo Scientific Chemicals Supplier No. 419590250
Only null left

CAS: 13517-24-3 | Na2SiO3·9H2O | 284.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13517-24-3
Molecular Formula Na2SiO3·9H2O
Molecular Weight (g/mol) 284.19
MDL Number MFCD00149175
InChI Key PHIQPXBZDGYJOG-UHFFFAOYSA-N
Synonym sodium metasilicate nonahydrate, unii-d8d44215lz, sodium silicate nonahydrate, silicic acid h2sio3 , disodium salt, nonahydrate, disodium oxosilanebis olate nonahydrate, 2na.so3i.9h2o, sodium meta-silicate nonahydrate, disodium oxosilanediolate nonahydrate, disodium dioxido oxo silane nonahydrate, sodium oxosilanebis olate-water 1/9
PubChem CID 61639
IUPAC Name disodium;dioxido(oxo)silane;nonahydrate
SMILES O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+]

Specifications

Melting Point 72.0°C
Color White
Physical Form Granules or Granular Powder
Quantity 25 g
Linear Formula Na2SiO3·9H2O
Merck Index 15, 8804
Solubility Information Solubility in water: soluble
Formula Weight 284.19
Percent Purity 44 to 47.5% total solids
Packaging Plastic bottle
Density 2.61
Chemical Name or Material Sodium metasilicate nonahydrate, 44 - 47.5% total solids
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

RUO – Research Use Only