missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Metasilicate, MP Biomedicals
$56.99 - $56.99
Chemical Identifiers
| CAS | 13517-24-3 |
|---|---|
| Molecular Formula | H18Na2O12Si |
| Molecular Weight (g/mol) | 284.197 |
| InChI Key | PHIQPXBZDGYJOG-UHFFFAOYSA-N |
| Synonym | sodium metasilicate nonahydrate, unii-d8d44215lz, sodium silicate nonahydrate, silicic acid h2sio3 , disodium salt, nonahydrate, disodium oxosilanebis olate nonahydrate, 2na.so3i.9h2o, sodium meta-silicate nonahydrate, disodium oxosilanediolate nonahydrate, disodium dioxido oxo silane nonahydrate, sodium oxosilanebis olate-water 1/9 |
| PubChem CID | 61639 |
| IUPAC Name | disodium;dioxido(oxo)silane;nonahydrate |
| SMILES | O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN19138280
|
MP Biomedicals Inc
0219138280 |
100 g |
Each for $56.99
|
|
|||||
Chemical Identifiers
| 13517-24-3 | |
| 284.197 | |
| sodium metasilicate nonahydrate, unii-d8d44215lz, sodium silicate nonahydrate, silicic acid h2sio3 , disodium salt, nonahydrate, disodium oxosilanebis olate nonahydrate, 2na.so3i.9h2o, sodium meta-silicate nonahydrate, disodium oxosilanediolate nonahydrate, disodium dioxido oxo silane nonahydrate, sodium oxosilanebis olate-water 1/9 | |
| disodium;dioxido(oxo)silane;nonahydrate |
| H18Na2O12Si | |
| PHIQPXBZDGYJOG-UHFFFAOYSA-N | |
| 61639 | |
| O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+] |
Specifications
| 13517-24-3 | |
| ≥97% | |
| sodium metasilicate nonahydrate, unii-d8d44215lz, sodium silicate nonahydrate, silicic acid h2sio3 , disodium salt, nonahydrate, disodium oxosilanebis olate nonahydrate, 2na.so3i.9h2o, sodium meta-silicate nonahydrate, disodium oxosilanediolate nonahydrate, disodium dioxido oxo silane nonahydrate, sodium oxosilanebis olate-water 1/9 | |
| O.O.O.O.O.O.O.O.O.[O-][Si](=O)[O-].[Na+].[Na+] | |
| 284.197 | |
| 284.2 | |
| Sodium Metasilicate |
| 100 g | |
| H18Na2O12Si | |
| PHIQPXBZDGYJOG-UHFFFAOYSA-N | |
| disodium;dioxido(oxo)silane;nonahydrate | |
| 61639 | |
| Reagent |
Safety and Handling
Recommended Storage : Room Temperature