missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hexametaphosphate (Powder/Laboratory), Fisher Chemical
Specifications
Specifications
| Melting Point | 640°C |
| Color | White |
| pH | 6.0 to 7.7 |
| Physical Form | Solid |
| Quantity | 500 g |
| Identification | Pass Test |
| Packaging | Glass Bottle |
| Density | 2.181 g/cm³ |
| Linear Formula | (NaPO3)n, where n ≈ 6 |
| Grade | Laboratory |
| Show More |