missing translation for 'onlineSavingsMsg'
Learn More

Sodium metaborate tetrahydrate, tech.

Catalog No. p-7050408
Change view
Click to view available options
Quantity:
1 kg
5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36736A1 1 kg
AA36736A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA36736A1 Supplier Thermo Scientific Chemicals Supplier No. 036736A1
Only null left

CAS: 10555-76-7 | BH8NaO6 | 137.86 g/mol

Sodium metaborate tetrahydrate is used In textile industry, it is used as additives, process aid and flame retardant. In chemical industry, it is used to make boric acid. Other uses include thickening agent, preservative and photographic chemical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10555-76-7
Molecular Formula BH8NaO6
Molecular Weight (g/mol) 137.86
MDL Number MFCD00149205
InChI Key JAKYJVJWXKRTSJ-UHFFFAOYSA-N
Synonym sodium metaborate tetrahydrate, sodium tetrahydrate oxoborinate, na.bo2.4h2o, boric acid, sodium salt, tetrahydrate
PubChem CID 23694267
IUPAC Name sodium;oxido(oxo)borane;tetrahydrate
SMILES O.O.O.O.[Na+].[O-]B=O

Specifications

Melting Point ∼53°C (decomposition)
Physical Form Crystalline
Quantity 1 kg
Solubility Information Soluble in water.
Formula Weight 137.86 (65.80 Anhydrous)
Grade Technical
Chemical Name or Material Sodium metaborate tetrahydrate
Hazard Category H319-H335-H361d
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c
EINECSNumber 231-891-6
RTECSNumber ED4640000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only