missing translation for 'onlineSavingsMsg'
Learn More

Sodium metaborate tetrahydrate, 98%

CAS: 10555-76-7 | BH8NaO6 | 137.86 g/mol

Supplier:  Thermo Scientific Chemicals A118030E

 View more versions of this product

Catalog No. AAA118030E

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium metaborate tetrahydrate
10555-76-7
BH8NaO6
sodium metaborate tetrahydrate, sodium tetrahydrate oxoborinate, na.bo2.4h2o, boric acid, sodium salt, tetrahydrate
O.O.O.O.[Na+].[O-]B=O
137.86
137.86 (65.80 Anhydrous)
Odorless
∽53°C (decomposition)
2500 g
MFCD00149205
JAKYJVJWXKRTSJ-UHFFFAOYSA-N
sodium;oxido(oxo)borane;tetrahydrate
23694267
98%

Chemical Identifiers

10555-76-7
137.86
JAKYJVJWXKRTSJ-UHFFFAOYSA-N
23694267
O.O.O.O.[Na+].[O-]B=O
BH8NaO6
MFCD00149205
sodium metaborate tetrahydrate, sodium tetrahydrate oxoborinate, na.bo2.4h2o, boric acid, sodium salt, tetrahydrate
sodium;oxido(oxo)borane;tetrahydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c

H319-H335-H361d

EINECSNumber : 231-891-6

RTECSNumber : ED4640000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only