missing translation for 'onlineSavingsMsg'
Learn More

Sodium metaborate tetrahydrate, 98.5%, extra pure

Catalog No. p-42097
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC211635000 500 g Plastic bottle
AC211630025 2.5 kg Plastic bottle
AC211630100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC211635000 Supplier Thermo Scientific Chemicals Supplier No. 211635000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10555-76-7 | BH8NaO6 | 137.86 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10555-76-7
Molecular Formula BH8NaO6
Molecular Weight (g/mol) 137.86
MDL Number MFCD00149205
InChI Key JAKYJVJWXKRTSJ-UHFFFAOYSA-N
Synonym sodium metaborate tetrahydrate, sodium tetrahydrate oxoborinate, na.bo2.4h2o, boric acid, sodium salt, tetrahydrate
PubChem CID 23694267
SMILES O.O.O.O.[Na+].[O-]B=O

Specifications

Melting Point 57.0°C
Color White
Physical Form Crystalline Powder and/or Crystals
Quantity 500 g
Assay Percent Range 98.5%
Linear Formula NaBO2·4H2O
Merck Index 10,8477
Solubility Information Solubility in water: 41.9% (20°C).
IUPAC Name sodium tetrahydrate oxoborinate
Formula Weight 137.86
Percent Purity 98.5%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Sodium metaborate tetrahydrate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical

RUO – Research Use Only