missing translation for 'onlineSavingsMsg'
Learn More

Sodium metabisulfite, SO{2} 58.5% min

Catalog No. AA3337536
Change view
Click to view available options
Quantity:
500 g
2 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3337536 500 g
AA33375A3 2 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3337536 Supplier Thermo Scientific Chemicals Supplier No. 03337536

CAS: 7681-57-4 | Na2O5S2 | 190.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
MDL Number MFCD00167602
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
Synonym sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
PubChem CID 656671
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O

Specifications

Melting Point >300°C
Color White to Yellow
pH 3.5 to 5.0
Physical Form Granules
Quantity 500 g
Merck Index 14,8638
Solubility Information Freely soluble in water, glycerol; Slightly soluble in alcohol
Formula Weight 190.1
Concentration or Composition (by Analyte or Components) SO2 58.5% min.
Odor Like rotten eggs
Sensitivity Air and moisture sensitive
Density 1.480 g/mL
Chemical Name or Material Sodium metabisulfite
Show More Show Less
Hazard Category H302-H318
Hazard Statement GHS H Statement
H318-H302
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P280-P301+P312-P305+P351+P338-P310-P314-P330-P501c
EINECSNumber 231-673-0
RTECSNumber UX8225000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only