missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Metabisulfite, puriss. p.a. ACS Reagent, Honeywell Fluka™
Puriss. p.a., ACS Reagent, Reag. Ph. Eur., dry, 98-100.5%
Supplier: Honeywell Chemicals 314481KG
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
| Sodium Metabisulfite | |
| 7681-57-4 | |
| 1 kg | |
| Na2S2O5 | |
| NONH for all modes of transport | |
| LDTLADDKFLAYJA-UHFFFAOYSA-L | |
| disodium (sulfinatooxy)sulfinate | |
| 656671 | |
| 98 to 100.5% |
| Reag. Ph. Eur. | |
| Dry | |
| Na2O5S2 | |
| MFCD00167602 | |
| sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis | |
| [Na+].[Na+].[O-]S(=O)OS([O-])=O | |
| 190.09 | |
| 190.11g/mol | |
| ACS/puriss. p.a. |
Chemical Identifiers
| 7681-57-4 | |
| 190.09 | |
| LDTLADDKFLAYJA-UHFFFAOYSA-L | |
| 656671 | |
| [Na+].[Na+].[O-]S(=O)OS([O-])=O |
| Na2O5S2 | |
| MFCD00167602 | |
| sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis | |
| disodium (sulfinatooxy)sulfinate |
Safety and Handling
P280-P301 + P312 + P330-P305 + P351 + P338 + P310
H302-H318
EINECSNumber : 231-673-0
RTECSNumber : UX8225000