missing translation for 'onlineSavingsMsg'
Learn More

Sodium metabisulfite, Puriss., meets analytical spec. of BP, FCC, NF, Ph. Eur., E223, dry, 97-100.5%, Honeywell Fluka™

Catalog No. p-7071811
Change view
Click to view available options
Quantity:
1 kg
4 x 5 kg
5 kg
6 x 1 kg
Catalog No. Quantity
6001992 1 kg
6001994 5 kg
60044810 6 x 1 kg
60044809 4 x 5 kg
4 options

Catalog No. 6001992

Supplier: Honeywell Chemicals 134591KG

Only null left

Puriss., meets analytical specification of BP, FCC, NF, Ph. Eur., E223, dry, 97-100.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
MDL Number MFCD00167602
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
Synonym sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
PubChem CID 656671
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O

Specifications

Physical Form Dry
Quantity 1 kg
Linear Formula Na2S2O5
UN Number NONH for all modes of transport
Formula Weight 190.11g/mol
Percent Purity 97 to 100.5%
Grade puriss.
Chemical Name or Material Sodium Metabisulfite, Meets analytical specification of Ph. Eur.BP, NF, FCC
Hazard Statement P280-P301 + P312 + P330-P305 + P351 + P338 + P310
Precautionary Statement H302-H318
EINECSNumber 231-673-0
RTECSNumber UX8225000