missing translation for 'onlineSavingsMsg'
Learn More

Sodium Metabisulfite (Granular/NF/FCC), Fisher Chemical™

Supplier:  Thermo Fisher Scientific FLS242212

 View more versions of this product

Catalog No. S242-212


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium Metabisulfite
White
4 to 6
2.5 kg
MFCD00167602
LDTLADDKFLAYJA-UHFFFAOYSA-L
disodium (sulfinatooxy)sulfinate
656671
65.0 to 67.4% (NF)
Pass Test (FCC), Pass Test (FCC, NF), Pass Test (NF)
1.4g/cm³
150°C
7681-57-4
Powder/Solid
Na2O5S2
sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
[Na+].[Na+].[O-]S(=O)OS([O-])=O
190.09
190.11
FCC/NF
Poly Bottle

Chemical Identifiers

7681-57-4
190.09
LDTLADDKFLAYJA-UHFFFAOYSA-L
656671
[Na+].[Na+].[O-]S(=O)OS([O-])=O
Na2O5S2
MFCD00167602
sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
disodium (sulfinatooxy)sulfinate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Harmful if swallowed. May cause skin and respiratory tract irritation. Contact with acids liberates toxic gas. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:0
Instability:1

SDS Documents

Documents

Promotions

Promotions