missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Metabisulfite, Anhydrous, Granular, Reagent, ACS, 97%, Spectrum™ Chemical
S1334, 7681-57-4, Na2S2O5
$394.33 - $6025.43
Chemical Identifiers
| CAS | 7681-57-4 |
|---|---|
| Molecular Formula | Na2O5S2 |
| Molecular Weight (g/mol) | 190.09 |
| MDL Number | MFCD00167602 |
| InChI Key | LDTLADDKFLAYJA-UHFFFAOYSA-L |
| IUPAC Name | disodium (sulfinatooxy)sulfinate |
| SMILES | [Na+].[Na+].[O-]S(=O)OS([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606638
|
Spectrum Chemical Mfg Cor
S1334500GM |
500 g | Poly Bottle |
Each for $394.33
|
|
||||
|
18606640
|
Spectrum Chemical Mfg Cor
S133425KG |
2.5 kg | Poly Bottle |
Each for $1,081.43
|
|
||||
|
18606639
|
Spectrum Chemical Mfg Cor
S1334500GMCS6 |
6 x 500 g | Poly Bottle |
Case of 6 Each for $1,712.30
|
|
||||
|
18606641
|
Spectrum Chemical Mfg Cor
S133425KGCS4 |
4 x 2.5 kg | Poly Bottle |
Case of 4 Each for $3,566.47
|
|
||||
|
18606642
|
Spectrum Chemical Mfg Cor
S133412KG |
12 kg | Poly Pail |
Each for $3,125.20
|
|
||||
|
18606643
|
Spectrum Chemical Mfg Cor
S133445KGBL |
45 kg | Fiber Drum |
Each for $6,025.43
|
|
||||
Description
Spectrum™ Chemical Sodium Metabisulfite, Anhydrous, Granular, Reagent, ACS , also known as disodium or metabisulfite, is typically used as an antioxidant, disinfectant or preservative agent. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.Chemical Identifiers
| 7681-57-4 | |
| 190.09 | |
| LDTLADDKFLAYJA-UHFFFAOYSA-L | |
| [Na+].[Na+].[O-]S(=O)OS([O-])=O |
| Na2O5S2 | |
| MFCD00167602 | |
| disodium (sulfinatooxy)sulfinate |
Specifications
| 7681-57-4 | |
| 500 g | |
| MFCD00167602 | |
| [Na+].[Na+].[O-]S(=O)OS([O-])=O | |
| 190.09 | |
| ACS |
| 1 | |
| Na2O5S2 | |
| LDTLADDKFLAYJA-UHFFFAOYSA-L | |
| disodium (sulfinatooxy)sulfinate | |
| 97% | |
| Poly Bottle |